Learn More
Sino Biological Influenza A H3N2 (A/Netherlands/178/1995) (RG145K) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40486V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Netherlands/178/1995)(RG145K)(H3N2)) hemagglutinin (AIU46036.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0973