Learn More
Sino Biological Influenza A H3N2 (A/Philippines/472/2002) (MDCK) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40487V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Philippines/472/2002 (MDCK)(H3N2)) hemagglutinin (AIU46063.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0979