Learn More
Sino Biological Influenza A H3N2 (A/Missouri/09/2014) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40494V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Missouri/09/2014(H3N2)) hemagglutinin (translated amino acids of EPI552125) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0995