Learn More
Sino Biological AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 Protein (His Tag)
Supplier:  Sino Biological 40496V08H100
Encompass_Preferred
A DNA sequence encoding the autographa californica nucleopolyhedrovirus (AcNPV)(strain E2) gp64 (NP_054158.1) (Met1-Thr481) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1003