Learn More
Sino Biological Influenza A H3N2 (A/Switzerland/9715293/2013) Hemagglutinin / HA0 Protein (full length)
Supplier:  Sino Biological 40497VNAB100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Switzerland/9715293/2013(H3N2)) hemagglutinin (translated amino acids of EPI541659) (Met1-Ile566), termed as HA0, was expressed.
Catalog No. 50-162-1010