Learn More
Sino Biological Influenza B (B/PHUKET/3073/2013) Hemagglutinin / HA0 Protein (full length)
Supplier:  Sino Biological 40498VNAB100
Encompass_Preferred
A DNA sequence encoding the full length of the Influenza virus B (B/PHUKET/3073/2013) hemagglutinin (translated amino acids of EPI529345) (Met1-Leu584), termed as HA, was expressed.
Catalog No. 50-162-1017