Learn More
Sino Biological Influenza A H3N2 (A/Switzerland/9715293/2013) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40499V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Switzerland/9715293/2013 (H3N2)) nucleoprotein (translated amino acids of EPI541652) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1019