Learn More
Jackson Immuno Research Labs DyLight 405-AffiniPure F(ab')₂ Goat Anti-Human IgG, Fcγ, 0.75 mg, blue-fluorescing, 400 nm excitation, 421 nm emission, affinity-purified, freeze-dried solid
SDP
Supplier:  Jackson Immuno Research Labs 109476170
Encompass_Preferred
DyLight 405-AffiniPure F(ab')₂ Goat Anti-Human IgG, Fcγ, 0.75 mg, blue-fluorescing, 400 nm excitation, 421 nm emission, affinity-purified, freeze-dried solid
Catalog No. NC1592036
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov