Learn More
METTLER TOLEDO 405-DPAS-SC-K8S pH Probe
Supplier:  METTLER TOLEDO 59903234
Encompass
405-DPAS is a pre-pressurized, gel-filled, low maintenance pH probe, specially developped for in-line measurements in biotech and food & beverage applications. It is designed for use in reactors, tanks, open basins and transfer lines under harsh chemical process conditions.
Catalog No. NC1011837