Learn More
Sino Biological Influenza B (B/PHUKET/3073/2013) Neuraminidase / NA Protein (His Tag)
Supplier:  Sino Biological 40502V07B100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/PHUKET/3073/2013) neuraminidase (translated amino acids of EPI529344) (Leu38-Leu466) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1021