Learn More
Sino Biological Influenza A H1N1 (A/Bel/1942) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40506V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Bel/1942 (H1N1)) hemagglutinin (ABD62843.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1023