Learn More
Sino Biological Influenza A H1N1 (A/Memphis/1/1987) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40509V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Memphis/1/1987 (H1N1)) hemagglutinin (ABQ44416.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1035