Learn More
Sino Biological Influenza A H5N8 (A/turkey/Germany-MV/R2472/2014) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40512V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/turkey/Germany-MV/R2472/2014 (H5N8)) hemagglutinin (translated amino acids of EPI544756) (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1045