Learn More
Sino Biological Influenza A H5N2 (A/chicken/Iowa/04-20/2015) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40518V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/chicken/Iowa/04-20/2015)(H5N2)) hemagglutinin (AKG52687.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1051