Learn More
Sino Biological SARS-CoV (strain WH20) Plpro / papain-like protease (His Tag)
Supplier:  Sino Biological 40524V08E100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV (strain WH20) papain-like protease (PLpro) (AAX16193.1) (Glu1541-Tyr1859) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1055