Learn More
Sino Biological Dengue virus (DENV) (type 1, strain US/Hawaii/1944) DENV-NS1 Protein (His Tag)
Supplier:  Sino Biological 40527V07H100
Encompass_Preferred
A DNA sequence encoding the Dengue virus (DENV) (type 1, strain US/Hawaii/1944) NS1 (ACF49259.1) (Asp776-Ala1127) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1058