Learn More
Sino Biological Dengue virus (DENV) (type 3, strain Philippines/H87/1956) NS1 Protein (His Tag)
Supplier:  Sino Biological 40528V07H100
Encompass_Preferred
A DNA sequence encoding the Dengue virus (DENV) (type 3, strain Philippines/H87/1956) NS1 (AAA99437.1) (Asp774-Ala1125) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1060