Learn More
Sino Biological Dengue virus (DENV) (type 4, strain Philippines/H241/1956) E / Envelope Protein (ECD, His Tag)
Supplier:  Sino Biological 40533V08B2100
Encompass_Preferred
A DNA sequence encoding the envelope protein of Dengue virus (DENV)(type 4, strain Philippines/H241/1956) (AAX48017.1) (Met280-Gly674) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1066