Learn More
Sino Biological Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E / Envelope protein (Domain III, Fc Tag)
Supplier:  Sino Biological 40543V02H100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) E / Envelope (domain III) (ALU33341.1) (Val593-Lys699) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1076