Learn More
Sino Biological Zika virus (ZIKV) (strain Zika SPH2015) M / Membrane protein (Fc Tag)
Supplier:  Sino Biological 40543V02H2100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) M / Membrane (ALU33341.1) (Ala216-Ser290) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1078