Learn More
Sino Biological Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E (Stem/anchor domain of flavivirus envelope glycoprotein E) protein (Fc Tag)
Supplier:  Sino Biological 40543V02H3100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) E_stem (Stem/anchor domain of flavivirus envelope glycoprotein E) (ALU33341.1) (Gly698-Ala794) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1081