Learn More
Sino Biological Zika virus (strain Zika SPH2015) E / Envelope protein (Domain III, His Tag)
Supplier:  Sino Biological 40543V08Y100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) E / Envelope (domain III) (ALU33341.1) (Val593-Lys699) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1094