Learn More
Sino Biological Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 protein (His Tag)
Supplier:  Sino Biological 40544V08B1100
Encompass_Preferred
A DNA sequence encoding the the Zika virus (strain Zika SPH2015) NS1 (ALU33341.1) (Val796-Leu1157) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1100