Learn More
Sino Biological Influenza A H5N6 (A/yunnan/0127/2015) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40557V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Yunnan/0127/2015 (H5N6)) hemagglutinin (AKS48023.1) (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1109