Learn More
Sino Biological Influenza A H1N1 (A/Michigan/45/2015) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40568V07H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Michigan/45/2015 (H1N1pdm09)) neuraminidase (translated amino acids of EPI859651) (His36-Lys469) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1113