Learn More
Ace Glass, Inc. Vacuum manifold, 405mm long, (4) stopcock ports with 24/40inner joints
SDP
Catalog No. 50976313
Encompass_Preferred
405mm long, single tube vacuum manifold featuring 4 stopcock ports with 24/40 inner joints. Distance between ports is 95mm which allows for use of flasks up to 3L.
Catalog No. 50-976-313 Supplier Ace Glass, Inc. Supplier No. 874533
405mm long, single tube vacuum manifold featuring 4 stopcock ports with 24/40 inner joints. Distance between ports is 95mm which allows for use of flasks up to 3L.