Learn More
Ace Glass, Inc. Vacuum manifold, 405mm long, (4) stopcock ports with #15o-ring joints
SDP
Catalog No. 50979844
Encompass_Preferred
405mm long, single tube vacuum manifold featuring 4 stopcock ports with #15 o-ring joints. Distance between ports is 95mm which allows for use of flasks up to 3L.
Catalog No. 50-979-844 Supplier Ace Glass, Inc. Supplier No. 874535
405mm long, single tube vacuum manifold featuring 4 stopcock ports with #15 o-ring joints. Distance between ports is 95mm which allows for use of flasks up to 3L.