Learn More
eMolecules​ 40949-94-8 | CALLERY? Potassium hexamethyldisilazane, 20% solution in tetrahydrofuran | Strem Chemicals | MFCD00010330 | 199.485 | C6H18KNSi2 | 0.000 | [K+].C[Si](C)(C)[N-][Si](C)(C)C | 500g | 689248445
Catalog No. 5000489246 Shop All eMolecules Inc. Products
Encompass_Preferred
CALLERY? Potassium hexamethyldisilazane, 20% solution in tetrahydrofuran | Strem Chemicals | 40949-94-8 | MFCD00010330 | 199.485 | C6H18KNSi2 | 0.000 | [K+].C[Si](C)(C)[N-][Si](C)(C)C | 500g | 689248445
Catalog No. 50-004-89246 Supplier eMolecules​ Supplier No. 689248445
CALLERY? Potassium hexamethyldisilazane, 20% solution in tetrahydrofuran | Strem Chemicals | 40949-94-8 | MFCD00010330 | 199.485 | C6H18KNSi2 | 0.000 | [K+].C[Si](C)(C)[N-][Si](C)(C)C | 500g | 689248445