Learn More
Mitegen 40K Cryogenic Freezer with CS200 Controller From IC Biomedical, 41,600 Vial Capacity, LN2 Liquid/Vapor Storage
SDP
Catalog No. NC3666381
Encompass
The 40K Cryogenic Freezer with CS200 Controller From IC Biomedical is a high-capacity LN2 system, storing up to 41,600 2ml vials at consistent near-LN2 temperatures (liquid or vapor). High efficiency and very low liquid nitrogen consumption ensure long-term sample viability. It features a smooth aluminum turntable for easy inventory access and a hinged, lockable lid with automatic defog. A stainless steel tabletop provides a work surface. The CS200 control system precisely monitors liquid levels and temperatures, offering data collection and robust sample security. An integral folding step and flexible fill hose enhance usability. This system is a high-performance solution for large-scale cryogenic sample management.
Catalog No. NC3666381 Supplier Mitegen Supplier No. TWLABS40KCS
Add to Cart
Add to Cart
Request a Quote
The 40K Cryogenic Freezer with CS200 Controller From IC Biomedical is a high-capacity LN2 system, storing up to 41,600 2ml vials at consistent near-LN2 temperatures (liquid or vapor). High efficiency and very low liquid nitrogen consumption ensure long-term sample viability. It features a smooth aluminum turntable for easy inventory access and a hinged, lockable lid with automatic defog. A stainless steel tabletop provides a work surface. The CS200 control system precisely monitors liquid levels and temperatures, offering data collection and robust sample security. An integral folding step and flexible fill hose enhance usability. This system is a high-performance solution for large-scale cryogenic sample management.