Learn More
C&G Containers 40 mL Clear Borosilicate Vial, 24-414 Thread Finish, Open Top Cap, 72 vials/case
SDP
Catalog No. NC1975909
Encompass_Preferred
40 mL Clear Borosilicate Glass Vial: Ideal for Volatile Organic Analysis (VOA), this authenticated and pre-cleaned vial ensures sample integrity. Featuring a 24-414 thread finish and a PTFE/silicone septum, it is designed for compatibility with various analytical instruments, minimizing contamination risks. The optimized vial shoulder prevents air bubble entrapment, making it a reliable choice for critical analytical applications.
Catalog No. NC1975909 Supplier C&G Containers Supplier No. LV79A000A01BZS1
40 mL Clear Borosilicate Glass Vial: Ideal for Volatile Organic Analysis (VOA), this authenticated and pre-cleaned vial ensures sample integrity. Featuring a 24-414 thread finish and a PTFE/silicone septum, it is designed for compatibility with various analytical instruments, minimizing contamination risks. The optimized vial shoulder prevents air bubble entrapment, making it a reliable choice for critical analytical applications.