Learn More
C&G Containers 40ML AMB GLASS VIAL CS/80
SDP
Catalog No. NC2669831
Encompass_Preferred
40ml Amber glass vial 24-414 with a white pp 2-piece hole cap with a Clear PTFE Septa, certified by cleaning protocol B1/Barcoded vials preserved with SULFURIC ACID (0.5ml 1:1) 80 vials/case.
Catalog No. NC2669831 Supplier C&G Containers Supplier No. LW54C100B51BZS1
40ml Amber glass vial 24-414 with a white pp 2-piece hole cap with a Clear PTFE Septa, certified by cleaning protocol B1/Barcoded vials preserved with SULFURIC ACID (0.5ml 1:1) 80 vials/case.