Learn More
C&G Containers 40ML AMB HCL 1ML 80PK
SDP
Catalog No. NC0661724
Encompass_Preferred
40ml Amber glass vial with 24-414 white PP 1-piece hole cap with sonically bonded PTFE Septa pre-cleaned and certified B1 Bar coded/(1ml) Hydrochloric Acid 1:1, 80 vials/rack
Catalog No. NC0661724 Supplier C&G Containers Supplier No. LW54H100C11BZS1
40ml Amber glass vial with 24-414 white PP 1-piece hole cap with sonically bonded PTFE Septa pre-cleaned and certified B1 Bar coded/(1ml) Hydrochloric Acid 1:1, 80 vials/rack