Learn More
C&G Containers C&G Containers 40mL Amber Vial, Certified B1 for Volatile Analytes, Type 1 Borosilicate Glass
SDP
Catalog No. NC3347956
Encompass_Preferred
The C&G Containers 40mL Amber Vial is a highly reliable Type 1 borosilicate glass container tailored for sensitive sample collection, ensuring minimal contamination for light-sensitive applications like Volatile Organic Analysis (VOA). Its secure open-top screw cap and certified precleaning enhance sample integrity, making it ideal for various organic analysis requirements. This vial stands out with its light protection and compatibility with industry-standard caps.
Catalog No. NC3347956 Supplier C&G Containers Supplier No. LW64A000A01BZS1
The C&G Containers 40mL Amber Vial is a highly reliable Type 1 borosilicate glass container tailored for sensitive sample collection, ensuring minimal contamination for light-sensitive applications like Volatile Organic Analysis (VOA). Its secure open-top screw cap and certified precleaning enhance sample integrity, making it ideal for various organic analysis requirements. This vial stands out with its light protection and compatibility with industry-standard caps.