Learn More
Analytical Sales & Services 40mL Amber Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners
Catalog No. 5000805472
Encompass
40mL Amber Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners
Catalog No. 50-008-05472 Supplier Analytical Sales & Services Supplier No. ADV376840
40mL Amber Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners