Learn More
C&G Containers 40 mL Amber Glass Vial, High Chemical Resistance, UV Protection, 24-414 Neck Finish
SDP
Catalog No. NC3591717
Encompass_Preferred
Introducing the 40 mL Amber Glass Vial, a reliable solution for scientific sample storage and protection. With its high-quality borosilicate glass construction, this vial offers excellent chemical resistance and UV protection for sensitive samples. Its compatible polypropylene screw cap ensures a secure, leak-proof seal. Ideal for environmental analysis, it meets strict EPA and ASTM standards, making it a trusted choice for various laboratory applications.
Catalog No. NC3591717 Supplier C&G Containers Supplier No. LAV040700005
Introducing the 40 mL Amber Glass Vial, a reliable solution for scientific sample storage and protection. With its high-quality borosilicate glass construction, this vial offers excellent chemical resistance and UV protection for sensitive samples. Its compatible polypropylene screw cap ensures a secure, leak-proof seal. Ideal for environmental analysis, it meets strict EPA and ASTM standards, making it a trusted choice for various laboratory applications.