Learn More
C&G Containers 40 mL Clear VOA Vial, Type 1 Borosilicate Glass, 24-414 White Open Top Cap, 80 vials/case
SDP
Catalog No. NC1922653
Encompass_Preferred
Introducing the 40 mL Clear VOA Vial from C & G CONTAINERS, engineered for precise Volatile Organic Compound Analysis. Made from durable Type 1 Borosilicate glass, this vial ensures reliable chemical resistance and clarity, essential for meticulous testing. Each vial comes with a 24-414 white cap featuring a bonded septum, effectively preventing contamination and sample evaporation. Packaged in cases of 80, it's ideal for high-demand laboratory environments.
Catalog No. NC1922653 Supplier C&G Containers Supplier No. LV53A020A01BZS1
Introducing the 40 mL Clear VOA Vial from C & G CONTAINERS, engineered for precise Volatile Organic Compound Analysis. Made from durable Type 1 Borosilicate glass, this vial ensures reliable chemical resistance and clarity, essential for meticulous testing. Each vial comes with a 24-414 white cap featuring a bonded septum, effectively preventing contamination and sample evaporation. Packaged in cases of 80, it's ideal for high-demand laboratory environments.