Learn More
Analytical Sales & Services 40mL Clear Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners
Catalog No. 5000806648
Encompass
Advantage™ Assembled Vial Kit. Contains: 3000 Class 40mL Clear Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners
Catalog No. 50-008-06648 Supplier Analytical Sales & Services Supplier No. ADV376740
Advantage™ Assembled Vial Kit. Contains: 3000 Class 40mL Clear Glass EPA/VOA Vials and Polypropylene Open Top Caps with Teflon/Silicone Liners