Learn More
Scientific Specialties Service Inc 40ML CLEAR TOC VIALS 72PK
SDP
Catalog No. NC2480047
Encompass_Preferred
0 mL, Clear, Low Level TOC Vials - Polypropylene Open Top Caps and Septrseal PTFE/Silicone Septa; Precleaned for Low Level TOC Analysis
Catalog No. NC2480047 Supplier Scientific Specialties Service Inc Supplier No. 276740TOCVAC
0 mL, Clear, Low Level TOC Vials - Polypropylene Open Top Caps and Septrseal PTFE/Silicone Septa; Precleaned for Low Level TOC Analysis