Learn More
ERA WATERS CORP 40 mL Low TOC autosampler vials, 80/cs
SDP
Catalog No. NC0861813
Encompass
Low TOC vials certified <10 ppb TOC, 40 mL Low TOC Vials (80/cs).
Catalog No. NC0861813 Supplier ERA WATERS CORP Supplier No. 25025
Low TOC vials certified <10 ppb TOC, 40 mL Low TOC Vials (80/cs).