Learn More
C&G Containers 40ML CLR GLS VL WHT/PLYCP 80PK
SDP
Catalog No. NC0607766
Encompass_Preferred
40ml FLINT GLASS VIAL 24-414 WITH WHITE PTFE THIN SEPTA D1 <10PPB TOC IN PINK ANTI-STATIC SLOTTED TRAY WITH DUST COVERS 80 VIALS/PACK
Catalog No. NC0607766 Supplier C&G Containers Supplier No. LFV040700005
40ml FLINT GLASS VIAL 24-414 WITH WHITE PTFE THIN SEPTA D1 <10PPB TOC IN PINK ANTI-STATIC SLOTTED TRAY WITH DUST COVERS 80 VIALS/PACK