Learn More
ERA WATERS CORP Low TOC vials certified <10 ppb TOC, 40 mL Low TOC Vials (80/cs).
SDP
Catalog No. NC1863139
Encompass
Low TOC vials certified <10 ppb TOC, 40 mL Low TOC Vials (80/cs).
Catalog No. NC1863139 Supplier ERA WATERS CORP Supplier No. 25025ERA
Low TOC vials certified <10 ppb TOC, 40 mL Low TOC Vials (80/cs).