Learn More
Ace Glass, Inc. 40mL porosity A extraction thimble
SDP
Catalog No. 50978883
Encompass_Preferred
40mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-978-883 Supplier Ace Glass, Inc. Supplier No. 681302
40mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.