Learn More
Ace Glass, Inc. 40mL porosity B extraction thimble
SDP
Catalog No. 50973431
Encompass_Preferred
40mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-431 Supplier Ace Glass, Inc. Supplier No. 681304
40mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.