Learn More
Ace Glass, Inc. 40mL porosity E extraction thimble
SDP
Catalog No. 50978884
Encompass_Preferred
40mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-978-884 Supplier Ace Glass, Inc. Supplier No. 681310
40mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.