Learn More
Alkali Scientific 40mL Precleaned VOA Vial, EPA Class 2, Clear Glass 27.5x95mm, 24 414 Cap, PTFE/Silicone Septa, Pre Assembled, 72/Pack
SDP
Catalog No. 5000662579
Encompass_Preferred
These 40mL Clear Glass VOA/TOC Vials are precleaned to EPA Class 2 standards (processed), ensuring high purity for volatile organic analysis (VOA), total organic carbon (TOC) testing, and other regulated environmental applications. Each 27.5 x 95mm vial is made from durable borosilicate glass and comes pre assembled with a 24 414 white open top polypropylene (PP) screw cap and a 22mm natural PTFE/white silicone septa (3.0mm thick) for excellent chemical resistance and airtight sealing. Designed for environmental labs, water testing, and analytical procedures, these precleaned glass vials minimize contamination and reduce preparation time. Supplied in a convenient 72 pack, they're ideal for use in EPA compliant sampling protocols. 40mL Clear Glass Vial, dimensions: 27.5 x 95mm Precleaned EPA Class 2, suitable for VOC, VOA, TOC analysis Optimized for use with automation Pre assembled with 24 414 PP screw cap and PTFE/silicone septa Septa: 22mm diameter, 3.0mm thick, chemically resistant
Catalog No. 50-006-62579 Supplier Alkali Scientific Supplier No. VOA040
These 40mL Clear Glass VOA/TOC Vials are precleaned to EPA Class 2 standards (processed), ensuring high purity for volatile organic analysis (VOA), total organic carbon (TOC) testing, and other regulated environmental applications. Each 27.5 x 95mm vial is made from durable borosilicate glass and comes pre assembled with a 24 414 white open top polypropylene (PP) screw cap and a 22mm natural PTFE/white silicone septa (3.0mm thick) for excellent chemical resistance and airtight sealing. Designed for environmental labs, water testing, and analytical procedures, these precleaned glass vials minimize contamination and reduce preparation time. Supplied in a convenient 72 pack, they're ideal for use in EPA compliant sampling protocols. 40mL Clear Glass Vial, dimensions: 27.5 x 95mm Precleaned EPA Class 2, suitable for VOC, VOA, TOC analysis Optimized for use with automation Pre assembled with 24 414 PP screw cap and PTFE/silicone septa Septa: 22mm diameter, 3.0mm thick, chemically resistant