Learn More
Lab Products Inc 40 mL Vial With PTFE Lined Cap, Pre-Cleaned, Certified For VOA Testing, 72 Vials/Case
Catalog No. NC9887145
Encompass_Preferred
Introducing the 40 mL Vial with PTFE Cap, designed for precision in laboratory analysis. Crafted from Type I borosilicate glass, this vial ensures sample integrity for VOLA and TOC testing. Its PTFE-lined silicone septum and screw cap provide secure closure, while being pre-cleaned and certified. With convenient packaging of 72 per case, it stands out for its reliability in sensitive scientific applications, meeting EPA guidelines.
Catalog No. NC9887145 Supplier Lab Products Inc Supplier No. 340C72TPC
Introducing the 40 mL Vial with PTFE Cap, designed for precision in laboratory analysis. Crafted from Type I borosilicate glass, this vial ensures sample integrity for VOLA and TOC testing. Its PTFE-lined silicone septum and screw cap provide secure closure, while being pre-cleaned and certified. With convenient packaging of 72 per case, it stands out for its reliability in sensitive scientific applications, meeting EPA guidelines.