Learn More
Azzota Corp 40mm 4-side clear Quartz 14ml
Catalog No. 5000887181
Encompass_Preferred
Four (4) sides clear Pathlength - 40mm Inside width - 10mm Volume - 14 0ml Dimensions - 45x12 5x42 5mm Type Q - Quartz (190nm-2500nm)
Catalog No. 50-008-87181 Supplier Azzota Corp Supplier No. Q207
Four (4) sides clear Pathlength - 40mm Inside width - 10mm Volume - 14 0ml Dimensions - 45x12 5x42 5mm Type Q - Quartz (190nm-2500nm)