Learn More
Bioptechs Inc 40mm German Desag 263 borosilicate glass coverslips, 500/cs
Catalog No. NC1408922
Encompass_Preferred
Package of 40mm German Desag 263 borosilicate glass coverslips, ideal for imaging. Used in the Bioptechs FCS2 and FCS3 Chamber systems.
Catalog No. NC1408922 Supplier Bioptechs Inc Supplier No. 40131303192
Package of 40mm German Desag 263 borosilicate glass coverslips, ideal for imaging. Used in the Bioptechs FCS2 and FCS3 Chamber systems.