Learn More
Ace Glass, Inc. 40MM GRADED SEALS 40MM GRADED SEALS
SDP
Catalog No. 50979785
Encompass
40mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-785 Supplier Ace Glass, Inc. Supplier No. 845548
40mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.