Learn More
Ace Glass, Inc. 40mm straight tubes with 145-174 micron fritted discs
SDP
Catalog No. 50974382
Encompass_Preferred
40mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-382 Supplier Ace Glass, Inc. Supplier No. 720532
40mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.